The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244847.1 | NP_001231776.1 | 4 - 5 | 310 | 66 |
NM_207392.2 | NP_997275.1 | 5 - 6 | 352 | 66 | |
GenBank mRNA | AY359007.1 | - | 5 - 6 | 275 | 66 |
BC130501.1 | - | 5 - 6 | 352 | 66 |