The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080476.2 | NP_001073945.1 | 2 - 3 | 84 | |
GenBank mRNA | AA588868.1 | - | 2 - 3 | 364 | 84 |