The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178569.3 | NP_848664.1 | 3 - 4 | 540 | 79 |
NR_122126.1 | 3 - 4 | 482 | 79 | ||
XM_011513975.2 | XP_011512277.1 | 3 - 4 | 700 | 79 | |
GenBank mRNA | AY249325.1 | - | 3 - 4 | 523 | 79 |
BC101608.1 | - | 3 - 4 | 443 | 79 | |
BC101634.1 | - | 3 - 4 | 443 | 79 | |
BC143638.1 | - | 3 - 4 | 385 | 79 |