Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CUL2
Entrez Gene ID
8453
Gene Name
cullin 2
Gene Aliases
-
Chromosome Location
Chr.10: 35008551 - 35126899 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011519743.1 XP_011518045.1 1 - 2 51 139

View Full Product Details