Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HEATR9
Entrez Gene ID
256957
Gene Name
HEAT repeat containing 9
Gene Aliases
C17orf66
Chromosome Location
Chr.17: 35854946 - 35868891 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321395.1 NP_001308324.1 14 - 15 1397 93
NM_152781.3 NP_689994.2 14 - 15 1517 93
NR_135630.1 14 - 15 1561 93
XM_005278233.2 XP_005278290.1 15 - 16 1476 93
XM_005278235.1 XP_005278292.1 15 - 16 1361 93
XM_006721802.3 XP_006721865.1 11 - 12 999 93
XM_011524586.2 XP_011522888.1 14 - 15 1460 93
GenBank mRNA AK057392.1 - 14 - 15 1517 93
AK301943.1 - 14 - 15 1377 93
BC033734.1 - 13 - 14 2151 93
BC132681.1 - 14 - 15 1453 93
BC144011.1 - 14 - 15 1497 93
DB090576.1 - 1 - 2 93

View Full Product Details