Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAMD9
Entrez Gene ID
54809
Gene Name
sterile alpha motif domain containing 9
Gene Aliases
C7orf5, DRIF1, NFTC, OEF1, OEF2
Chromosome Location
Chr.7: 93099513 - 93118023 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017654.3 NP_060124.2 2 - 3 263 128
GenBank mRNA AB095925.2 - 2 - 3 197 128
AF445355.1 - 2 - 3 263 128
AF453311.3 - 2 - 3 227 128
BC065769.1 - 2 - 3 253 128
BC132773.1 - 1 - 2 93 128
BC132775.1 - 1 - 2 93 128
BC150249.1 - 2 - 3 218 128
BG615970.1 - 2 - 3 250 128
BX647072.1 - 2 - 3 219 128

View Full Product Details