The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322831.1 | NP_001309760.1 | 19 - 20 | 2566 | 76 |
NM_001322834.1 | NP_001309763.1 | 19 - 20 | 2566 | 76 | |
NM_031913.4 | NP_114119.2 | 19 - 20 | 2566 | 76 | |
NR_136409.1 | 19 - 20 | 2800 | 76 | ||
XM_017007307.1 | XP_016862796.1 | 18 - 19 | 2018 | 76 | |
GenBank mRNA | AK125080.1 | - | 19 - 20 | 2523 | 76 |
BC037292.1 | - | 18 - 19 | 3524 | 76 | |
BQ435892.1 | - | 1 - 2 | 296 | 76 | |
DQ993202.1 | - | 19 - 20 | 2340 | 76 |