The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130009.2 | NP_001123481.2 | 3 - 4 | 551 | 111 |
NM_182625.4 | NP_872431.4 | 3 - 4 | 475 | 111 | |
XM_005262613.4 | XP_005262670.1 | 3 - 4 | 561 | 111 | |
XM_006712005.3 | XP_006712068.1 | 3 - 4 | 458 | 111 | |
XM_011532820.2 | XP_011531122.1 | 3 - 4 | 478 | 111 | |
XM_011532821.2 | XP_011531123.1 | 3 - 4 | 568 | 111 | |
XM_011532822.2 | XP_011531124.1 | 3 - 4 | 448 | 111 | |
GenBank mRNA | AK131387.1 | - | 3 - 4 | 423 | 111 |
BC117204.1 | - | 2 - 3 | 373 | 111 | |
BC117206.1 | - | 2 - 3 | 373 | 111 | |
CR936783.1 | - | 3 - 4 | 561 | 111 | |
DA317583.1 | - | 3 - 4 | 475 | 111 |