The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013623.2 | NP_001013645.1 | 2 - 3 | 195 | 94 |
GenBank mRNA | BC146950.1 | - | 2 - 3 | 195 | 94 |
BC146969.1 | - | 2 - 3 | 195 | 94 | |
BC171862.1 | - | 3 - 4 | 278 | 94 |