Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GKN2
Entrez Gene ID
200504
Gene Name
gastrokine 2
Gene Aliases
BRICD1B, GDDR, PRO813, TFIZ1, VLTI465
Chromosome Location
Chr.2: 68945232 - 68952970 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182536.2 NP_872342.2 2 - 3 170 66
GenBank mRNA AF494509.2 - 2 - 3 170 66
AJ966788.1 - 2 - 3 91 66
AY358664.1 - 2 - 3 145 66
AY943908.1 - 2 - 3 93 66

View Full Product Details