The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282430.1 | NP_001269359.1 | 1 - 2 | 226 | 107 |
GenBank mRNA | AY203953.1 | - | 1 - 2 | 385 | 107 |
BC150517.1 | - | 1 - 2 | 210 | 107 |