Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTXN3
Entrez Gene ID
613212
Gene Name
cortexin 3
Gene Aliases
KABE
Chromosome Location
Chr.5: 127649021 - 127658630 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001048252.2 NP_001041717.1 2 - 3 476 74
NM_001127385.1 NP_001120857.1 2 - 3 285 74
GenBank mRNA AB219764.1 - 2 - 3 453 74
AB219832.1 - 2 - 3 251 74
BC071790.1 - 2 - 3 249 74

View Full Product Details