The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136482.1 | NP_001129954.1 | 5 - 6 | 586 | 122 |
XM_005259846.4 | XP_005259903.1 | 6 - 7 | 932 | 122 | |
XM_005259847.4 | XP_005259904.1 | 6 - 7 | 774 | 122 | |
GenBank mRNA | BC157863.1 | - | 5 - 6 | 586 | 122 |