Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGBL2
Entrez Gene ID
79841
Gene Name
ATP/GTP binding protein like 2
Gene Aliases
CCP2
Chromosome Location
Chr.11: 47659168 - 47715397 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024783.3 NP_079059.2 4 - 5 535 83
XM_005253138.4 XP_005253195.1 3 - 4 980 83
XM_005253139.4 XP_005253196.1 3 - 4 980 83
XM_005253140.4 XP_005253197.1 3 - 4 980 83
XM_005253141.4 XP_005253198.1 3 - 4 980 83
XM_011520372.2 XP_011518674.1 4 - 5 889 83
XM_011520373.2 XP_011518675.1 3 - 4 980 83
XM_011520374.2 XP_011518676.1 3 - 4 980 83
XM_011520376.2 XP_011518678.1 4 - 5 1185 83
XM_011520381.2 XP_011518683.1 3 - 4 980 83
XM_017018331.1 XP_016873820.1 3 - 4 980 83
XM_017018333.1 XP_016873822.1 3 - 4 980 83
GenBank mRNA AK303066.1 - 4 - 5 488 83
BC036234.1 - 4 - 5 519 83

View Full Product Details