Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CORO1A
Entrez Gene ID
11151
Gene Name
coronin 1A
Gene Aliases
CLABP, CLIPINA, HCORO1, IMD8, TACO, p57
Chromosome Location
Chr.16: 30183410 - 30189076 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193333.2 NP_001180262.1 11 - 12 1715 121
NM_007074.3 NP_009005.1 10 - 11 1597 121
XM_011545714.2 XP_011544016.1 10 - 11 1410 121
GenBank mRNA AB209221.1 - 4 - 5 3981 121
AF495470.1 - 10 - 11 1402 121
AK096332.1 - 9 - 10 2592 121
AK314714.1 - 11 - 12 1516 121
BC110374.1 - 10 - 11 1414 121
BC126385.1 - 10 - 11 1301 121
BC126387.1 - 10 - 11 1301 121
D44497.1 - 10 - 11 1380 121
U34690.1 - 10 - 11 1372 121
X89109.1 - 10 - 11 1373 121

View Full Product Details