The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030928.3 | NP_112190.2 | 8 - 9 | 1332 | 82 |
GenBank mRNA | AB053172.1 | - | 8 - 9 | 1332 | 82 |
AF070552.1 | - | 6 - 7 | 871 | 82 | |
AF321125.1 | - | 8 - 9 | 1303 | 82 | |
BC000137.2 | - | 8 - 9 | 1294 | 82 | |
BC008676.1 | - | 8 - 9 | 1170 | 82 | |
BC008860.2 | - | 8 - 9 | 1225 | 82 | |
BC009410.1 | - | 8 - 9 | 1288 | 82 | |
BC014202.2 | - | 4 - 5 | 593 | 82 | |
BC021126.1 | - | 6 - 7 | 864 | 82 | |
BC049205.1 | - | 6 - 7 | 936 | 82 |