The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301772.1 | NP_001288701.1 | 1 - 2 | 125 | 147 |
NM_178172.5 | NP_835466.2 | 1 - 2 | 125 | 147 | |
GenBank mRNA | AY245915.1 | - | 1 - 2 | 147 | |
BC035810.1 | - | 1 - 2 | 147 | ||
BC063857.1 | - | 1 - 2 | 53 | 147 | |
HY047963.1 | - | 1 - 2 | 153 |