Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCDC5
Entrez Gene ID
100506627
Gene Name
doublecortin domain containing 5
Gene Aliases
-
Chromosome Location
Chr.11: 30863603 - 30992686 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AW340116.1 - 2 - 3 82
BX116244.1 - 1 - 2 80 82

View Full Product Details