Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BDNF-AS
Entrez Gene ID
497258
Gene Name
BDNF antisense RNA
Gene Aliases
ANTI-BDNF, BDNF, BDNF-AS1, BDNFOS, NCRNA00049
Chromosome Location
Chr.11: 27506852 - 27698171 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_002832.2 5 - 6 597 100
GenBank mRNA AY054403.4 - 5 - 6 627 100

View Full Product Details