The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288607.1 | NP_001275536.1 | 4 - 5 | 373 | 60 |
NM_001288608.1 | NP_001275537.1 | 4 - 5 | 373 | 60 | |
NM_019032.5 | NP_061905.2 | 4 - 5 | 373 | 60 | |
NM_025008.4 | NP_079284.2 | 4 - 5 | 373 | 60 | |
XM_011509644.2 | XP_011507946.1 | 3 - 4 | 335 | 60 | |
XM_011509645.2 | XP_011507947.1 | 3 - 4 | 332 | 60 | |
XM_011509648.2 | XP_011507950.1 | 3 - 4 | 289 | 60 | |
XM_011509649.2 | XP_011507951.1 | 3 - 4 | 343 | 60 | |
XM_011509650.2 | XP_011507952.1 | 3 - 4 | 343 | 60 | |
XM_017001506.1 | XP_016856995.1 | 4 - 5 | 744 | 60 | |
GenBank mRNA | AL513944.3 | - | 4 - 5 | 320 | 60 |
AY358122.1 | - | 3 - 4 | 187 | 60 | |
BC140800.1 | - | 3 - 4 | 225 | 60 | |
BC144581.1 | - | 3 - 4 | 225 | 60 | |
BM148104.1 | - | 1 - 2 | 81 | 60 | |
CB988088.1 | - | 4 - 5 | 361 | 60 |