The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113528.1 | NP_001107000.1 | 1 - 2 | 199 | 113 |
NM_001297775.1 | NP_001284704.1 | 1 - 2 | 199 | 113 | |
NM_152636.2 | NP_689849.2 | 1 - 2 | 199 | 113 | |
XM_011519935.2 | XP_011518237.1 | 1 - 2 | 197 | 113 | |
XM_011519940.2 | XP_011518242.1 | 1 - 2 | 199 | 113 | |
XM_011519941.2 | XP_011518243.1 | 1 - 2 | 199 | 113 | |
XM_017017298.1 | XP_016872787.1 | 1 - 2 | 197 | 113 | |
XM_017017301.1 | XP_016872790.1 | 1 - 2 | 199 | 113 | |
XM_017017303.1 | XP_016872792.1 | 1 - 2 | 197 | 113 | |
GenBank mRNA | DA800481.1 | - | 1 - 2 | 199 | 113 |
DB458698.1 | - | 1 - 2 | 202 | 113 |