Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANO6
Entrez Gene ID
196527
Gene Name
anoctamin 6
Gene Aliases
BDPLT7, SCTS, TMEM16F
Chromosome Location
Chr.12: 45215987 - 45440404 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204803.1 NP_001191732.1 1 - 2 404 77
GenBank mRNA BC136445.1 - 1 - 2 97 77

View Full Product Details