The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017637.5 | NP_060107.3 | 2 - 3 | 272 | 153 |
XM_017014816.1 | XP_016870305.1 | 3 - 4 | 2498 | 153 | |
XM_017014817.1 | XP_016870306.1 | 2 - 3 | 647 | 153 | |
XM_017014818.1 | XP_016870307.1 | 2 - 3 | 646 | 153 | |
XM_017014819.1 | XP_016870308.1 | 2 - 3 | 646 | 153 | |
XM_017014820.1 | XP_016870309.1 | 2 - 3 | 646 | 153 | |
XM_017014821.1 | XP_016870310.1 | 2 - 3 | 644 | 153 | |
XM_017014825.1 | XP_016870314.1 | 2 - 3 | 645 | 153 | |
XM_017014826.1 | XP_016870315.1 | 2 - 3 | 645 | 153 | |
XM_017014827.1 | XP_016870316.1 | 2 - 3 | 645 | 153 | |
XM_017014828.1 | XP_016870317.1 | 2 - 3 | 644 | 153 | |
GenBank mRNA | AK131398.1 | - | 2 - 3 | 154 | 153 |
DA494074.1 | - | 2 - 3 | 156 | 153 | |
DQ884933.1 | - | 2 - 3 | 463 | 153 | |
DQ884938.1 | - | 2 - 3 | 151 | 153 | |
DQ884939.1 | - | 3 - 4 | 258 | 153 | |
DQ884941.1 | - | 2 - 3 | 151 | 153 |