Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCDC5
Entrez Gene ID
100506627
Gene Name
doublecortin domain containing 5
Gene Aliases
-
Chromosome Location
Chr.11: 30863603 - 30992686 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020869.3 NP_065920.2 18 - 19 2857 106
GenBank mRNA AB040926.1 - 9 - 10 2869 106
BX117620.1 - 2 - 3 182 106
BX647348.1 - 17 - 18 4133 106

View Full Product Details