The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030962.3 | NP_112224.1 | 3 - 4 | 414 | 91 |
XM_005253154.4 | XP_005253211.1 | 3 - 4 | 423 | 91 | |
XM_005253155.4 | XP_005253212.1 | 3 - 4 | 420 | 91 | |
XM_011520394.2 | XP_011518696.1 | 3 - 4 | 420 | 91 | |
XM_011520395.2 | XP_011518697.1 | 3 - 4 | 423 | 91 | |
XM_011520396.2 | XP_011518698.1 | 3 - 4 | 423 | 91 | |
XM_017018372.1 | XP_016873861.1 | 2 - 3 | 1712 | 91 | |
XM_017018373.1 | XP_016873862.1 | 3 - 4 | 342 | 91 | |
XM_017018374.1 | XP_016873863.1 | 3 - 4 | 423 | 91 | |
XM_017018375.1 | XP_016873864.1 | 3 - 4 | 423 | 91 | |
XM_017018376.1 | XP_016873865.1 | 3 - 4 | 423 | 91 | |
XM_017018377.1 | XP_016873866.1 | 3 - 4 | 423 | 91 | |
GenBank mRNA | AB051553.1 | - | 2 - 3 | 191 | 91 |
AY234241.1 | - | 3 - 4 | 380 | 91 | |
BC101466.1 | - | 3 - 4 | 373 | 91 | |
CT000365.1 | - | 3 - 4 | 396 | 91 | |
DA862574.1 | - | 3 - 4 | 414 | 91 |