The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031735.2 | NP_001026905.2 | 2 - 3 | 375 | 84 |
NM_001039846.1 | NP_001034935.1 | 2 - 3 | 375 | 84 | |
XM_005259480.3 | XP_005259537.2 | 2 - 3 | 396 | 84 | |
GenBank mRNA | AK294083.1 | - | 2 - 3 | 246 | 84 |
AK301957.1 | - | 2 - 3 | 329 | 84 | |
AY358444.1 | - | 2 - 3 | 375 | 84 | |
AY753639.1 | - | 2 - 3 | 334 | 84 | |
BC010443.1 | - | 2 - 3 | 314 | 84 | |
BC014609.2 | - | 2 - 3 | 329 | 84 | |
BC040964.1 | - | 2 - 3 | 316 | 84 | |
BI827974.1 | - | 2 - 3 | 231 | 83 | |
BX378854.2 | - | 2 - 3 | 375 | 84 | |
DQ196309.1 | - | 2 - 3 | 362 | 84 | |
DQ196310.1 | - | 2 - 3 | 362 | 84 | |
DQ424900.1 | - | 2 - 3 | 298 | 84 |