The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198471.2 | NP_940873.2 | 10 - 11 | 2451 | 90 |
XM_006722718.3 | XP_006722781.1 | 10 - 11 | 2570 | 90 | |
XM_011527884.2 | XP_011526186.1 | 10 - 11 | 2827 | 90 | |
XM_017026563.1 | XP_016882052.1 | 9 - 10 | 2587 | 90 | |
GenBank mRNA | AB284126.1 | - | 10 - 11 | 2451 | 90 |
AK128815.1 | - | 10 - 11 | 2451 | 90 | |
BC066124.1 | - | 10 - 11 | 2451 | 90 |