Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD16
Entrez Gene ID
54522
Gene Name
ankyrin repeat domain 16
Gene Aliases
-
Chromosome Location
Chr.10: 5861726 - 5889897 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009941.2 NP_001009941.1 5 - 6 79
NM_019046.2 NP_061919.1 5 - 6 79
GenBank mRNA AA431197.1 - 2 - 3 296 79
AL137614.1 - 2 - 3 79
BC062346.1 - 5 - 6 79

View Full Product Details