The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244856.1 | NP_001231785.1 | 6 - 7 | 517 | 104 |
NM_198492.3 | NP_940894.1 | 6 - 7 | 553 | 104 | |
GenBank mRNA | AY203939.1 | - | 6 - 7 | 474 | 104 |
AY358431.1 | - | 7 - 8 | 550 | 104 | |
AY423706.1 | - | 6 - 7 | 553 | 104 | |
BC039679.1 | - | 2 - 3 | 140 | 104 | |
BC093691.1 | - | 6 - 7 | 507 | 104 | |
BC093693.1 | - | 6 - 7 | 507 | 104 | |
BC143309.1 | - | 6 - 7 | 471 | 104 |