Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TTC24
Entrez Gene ID
164118
Gene Name
tetratricopeptide repeat domain 24
Gene Aliases
-
Chromosome Location
Chr.1: 156579727 - 156587717 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105669.2 NP_001099139.2 7 - 8 1384 83
XM_011509253.2 XP_011507555.1 6 - 7 766 83
XM_011509254.2 XP_011507556.1 6 - 7 763 83
XM_011509255.2 XP_011507557.1 6 - 7 766 83
XM_017000497.1 XP_016855986.1 4 - 5 388 83
GenBank mRNA BQ216668.1 - 5 - 6 648 83
BX414790.2 - 4 - 5 530 83

View Full Product Details