Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FTCDNL1
Entrez Gene ID
348751
Gene Name
formiminotransferase cyclodeaminase N-terminal like
Gene Aliases
FONG
Chromosome Location
Chr.2: 199760536 - 199851173 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_034096.1 1 - 2 427 74
GenBank mRNA AB568489.1 - 1 - 2 374 74

View Full Product Details