Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GYG2
Entrez Gene ID
8908
Gene Name
glycogenin 2
Gene Aliases
GN-2, GN2
Chromosome Location
Chr.X: 2828712 - 2882820 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079855.1 NP_001073324.1 3 - 4 431 105
NM_001184702.1 NP_001171631.1 3 - 4 431 105
NM_001184703.1 NP_001171632.1 4 - 5 524 105
NM_003918.2 NP_003909.2 4 - 5 524 105
XM_011545600.2 XP_011543902.1 3 - 4 362 105
XM_017029927.1 XP_016885416.1 4 - 5 474 105
XM_017029928.1 XP_016885417.1 4 - 5 449 105
XM_017029929.1 XP_016885418.1 4 - 5 634 105
XM_017029930.1 XP_016885419.1 3 - 4 339 105
XM_017029931.1 XP_016885420.1 3 - 4 362 105
GenBank mRNA AK292496.1 - 3 - 4 339 105
BC023152.1 - 3 - 4 298 105
DQ424901.1 - 2 - 3 242 105
U94362.1 - 4 - 5 525 105
U94363.1 - 3 - 4 305 105

View Full Product Details