Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYT15
Entrez Gene ID
83849
Gene Name
synaptotagmin 15
Gene Aliases
CHR10SYT, sytXV
Chromosome Location
Chr.10: 46578434 - 46595170 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031912.4 NP_114118.2 5 - 6 976 69
NM_181519.2 NP_852660.1 5 - 6 976 69
XM_006718011.1 XP_006718074.1 5 - 6 976 69
XM_017016750.1 XP_016872239.1 5 - 6 986 69
XM_017016751.1 XP_016872240.1 5 - 6 976 69
XM_017016752.1 XP_016872241.1 5 - 6 976 69
GenBank mRNA AB109022.1 - 4 - 5 823 69
AB109023.1 - 4 - 5 823 69
AI971263.1 - 4 - 5 69
AJ303363.1 - 2 - 3 193 69
AK127436.1 - 4 - 5 1570 69
AK131036.1 - 5 - 6 976 69
AK290186.1 - 5 - 6 890 69
AK293632.1 - 5 - 6 654 69
BC139914.1 - 5 - 6 859 69

Gene Symbol
LOC102724488
Entrez Gene ID
102724488
Gene Name
synaptotagmin-15
Gene Aliases
-
Chromosome Location
Chr.10: 47745306 - 47763592 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006718093.3 XP_006718156.1 4 - 5 1570 69
XM_006718097.3 XP_006718160.1 5 - 6 968 69
XM_011540432.2 XP_011538734.1 4 - 5 1591 69
XM_011540434.2 XP_011538736.1 4 - 5 1591 69
XM_011540435.2 XP_011538737.1 4 - 5 1591 69
XM_011540436.2 XP_011538738.1 4 - 5 1591 69
XM_011540437.1 XP_011538739.1 4 - 5 1591 69
XM_011540438.2 XP_011538740.1 4 - 5 1591 69
XM_017017020.1 XP_016872509.1 5 - 6 986 69
XM_017017021.1 XP_016872510.1 5 - 6 968 69
XM_017017022.1 XP_016872511.1 4 - 5 1570 69
XM_017017023.1 XP_016872512.1 5 - 6 968 69
XM_017017024.1 XP_016872513.1 3 - 4 561 69

View Full Product Details