The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001115007.2 | NP_001108479.1 | 2 - 3 | 202 | 60 |
NM_001115008.2 | NP_001108480.1 | 2 - 3 | 403 | 60 | |
NM_001288997.1 | NP_001275926.1 | 2 - 3 | 140 | 60 | |
NR_110254.1 | 2 - 3 | 403 | 60 | ||
XM_017007730.1 | XP_016863219.1 | 2 - 3 | 397 | 60 | |
GenBank mRNA | AB111889.1 | - | 2 - 3 | 390 | 60 |
AK292769.1 | - | 2 - 3 | 150 | 60 | |
BX537919.1 | - | 2 - 3 | 300 | 60 | |
BX640657.1 | - | 2 - 3 | 129 | 60 |