The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173593.3 | NP_775864.3 | 3 - 4 | 354 | 83 |
GenBank mRNA | AB089940.1 | - | 3 - 4 | 354 | 83 |
CN400443.1 | - | 3 - 4 | 261 | 83 |