The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012454.5 | NP_001012457.3 | 4 - 5 | 860 | 81 |
NM_001128926.3 | NP_001122398.1 | 4 - 5 | 833 | 81 | |
NM_001290254.1 | NP_001277183.1 | 5 - 6 | 901 | 81 | |
NM_001290255.1 | NP_001277184.1 | 5 - 6 | 892 | 81 | |
XM_011516128.2 | XP_011514430.1 | 4 - 5 | 842 | 81 | |
XM_011516129.2 | XP_011514431.1 | 4 - 5 | 842 | 81 | |
XM_011516130.2 | XP_011514432.1 | 4 - 5 | 842 | 81 | |
XM_011516131.2 | XP_011514433.1 | 4 - 5 | 842 | 81 | |
XM_011516132.2 | XP_011514434.1 | 4 - 5 | 815 | 81 | |
XM_011516134.2 | XP_011514436.1 | 5 - 6 | 696 | 81 | |
XM_011516135.2 | XP_011514437.1 | 4 - 5 | 1198 | 81 | |
XM_011516136.2 | XP_011514438.1 | 4 - 5 | 1210 | 81 | |
XM_017012147.1 | XP_016867636.1 | 5 - 6 | 892 | 81 | |
GenBank mRNA | BC047310.1 | - | 4 - 5 | 832 | 81 |
BC066973.1 | - | 4 - 5 | 834 | 81 | |
BC105729.1 | - | 4 - 5 | 806 | 81 | |
BC105730.1 | - | 4 - 5 | 806 | 81 | |
BC105731.1 | - | 4 - 5 | 806 | 81 |