Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C3orf67
Entrez Gene ID
200844
Gene Name
chromosome 3 open reading frame 67
Gene Aliases
-
Chromosome Location
Chr.3: 58716415 - 59050078 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK128200.1 - 5 - 6 649 116

View Full Product Details