The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135219.1 | NP_001128691.1 | 6 - 7 | 637 | 74 |
XM_017014279.1 | XP_016869768.1 | 6 - 7 | 1798 | 74 | |
GenBank mRNA | BE881330.1 | - | 3 - 4 | 220 | 74 |