Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 81706053-81706636 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HGF
Entrez Gene ID
3082
Gene Name
hepatocyte growth factor
Gene Aliases
DFNB39, F-TCF, HGFB, HPTA, SF
UniGene
Hs.396530
Gene Chromosome Location
Chr.7: 81699006-81770438 on Build GRCh38
Transcripts
NM_000601.5 NM_001010932.2 XM_006715956.2 XM_011516115.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3030136 c.1711G>A p.V571I Chr.7: 81706333-81706333 on Build GRCh38
4529834 c.1646G>A p.G549E Chr.7: 81706398-81706398 on Build GRCh38
3431781 c.1617-2A>T p.? Chr.7: 81706429-81706429 on Build GRCh38
4529927 c.1654G>A p.D552N Chr.7: 81706390-81706390 on Build GRCh38
6937190 c.1676A>G p.E559G Chr.7: 81706368-81706368 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144086354 Chr.7: 81706370-81706370 on Build GRCh38
rs945603247 Chr.7: 81706210-81706210 on Build GRCh38
rs759934379 Chr.7: 81706454-81706454 on Build GRCh38
rs779933934 Chr.7: 81706381-81706381 on Build GRCh38
rs763167149 Chr.7: 81706319-81706319 on Build GRCh38
rs1052238811 Chr.7: 81706229-81706229 on Build GRCh38
rs773523345 Chr.7: 81706442-81706442 on Build GRCh38
rs201395847 Chr.7: 81706298-81706298 on Build GRCh38
rs970812279 Chr.7: 81706548-81706548 on Build GRCh38
rs946466823 Chr.7: 81706607-81706607 on Build GRCh38
rs952789397 Chr.7: 81706115-81706115 on Build GRCh38
rs984089630 Chr.7: 81706140-81706140 on Build GRCh38
rs769438908 Chr.7: 81706417-81706417 on Build GRCh38
rs5745750 Chr.7: 81706602-81706602 on Build GRCh38
rs547508089 Chr.7: 81706087-81706087 on Build GRCh38
rs560618119 Chr.7: 81706550-81706550 on Build GRCh38
rs576420401 Chr.7: 81706243-81706243 on Build GRCh38
rs935150791 Chr.7: 81706224-81706224 on Build GRCh38
rs763545388 Chr.7: 81706445-81706445 on Build GRCh38
rs1023624106 Chr.7: 81706532-81706532 on Build GRCh38
rs752976909 Chr.7: 81706462-81706462 on Build GRCh38
rs192078086 Chr.7: 81706395-81706395 on Build GRCh38
rs373049064 Chr.7: 81706281-81706281 on Build GRCh38
rs768245636 Chr.7: 81706250-81706250 on Build GRCh38
rs915519509 Chr.7: 81706591-81706591 on Build GRCh38
rs527424302 Chr.7: 81706220-81706220 on Build GRCh38
rs927881141 Chr.7: 81706555-81706555 on Build GRCh38
rs956838254 Chr.7: 81706578-81706578 on Build GRCh38
rs752743857 Chr.7: 81706346-81706346 on Build GRCh38
rs5745751 Chr.7: 81706581-81706581 on Build GRCh38
rs778704152 Chr.7: 81706369-81706369 on Build GRCh38
rs1041325785 Chr.7: 81706426-81706426 on Build GRCh38
rs771151236 Chr.7: 81706447-81706447 on Build GRCh38
rs773824390 Chr.7: 81706313-81706313 on Build GRCh38
rs752861058 Chr.7: 81706392-81706392 on Build GRCh38
rs1005681225 Chr.7: 81706517-81706517 on Build GRCh38
rs370757602 Chr.7: 81706372-81706372 on Build GRCh38
rs759776706 Chr.7: 81706336-81706336 on Build GRCh38
rs376043671 Chr.7: 81706357-81706357 on Build GRCh38
rs946475322 Chr.7: 81706307-81706307 on Build GRCh38
rs377268390 Chr.7: 81706466-81706466 on Build GRCh38
rs569479475 Chr.7: 81706145-81706145 on Build GRCh38
rs952766451 Chr.7: 81706507-81706507 on Build GRCh38
rs989738688 Chr.7: 81706176-81706176 on Build GRCh38
rs559990458 Chr.7: 81706359-81706359 on Build GRCh38
rs74658055 Chr.7: 81706461-81706461 on Build GRCh38
rs766769838 Chr.7: 81706329-81706329 on Build GRCh38
rs938202364 Chr.7: 81706157-81706157 on Build GRCh38
rs200799622 Chr.7: 81706485-81706485 on Build GRCh38
rs927774111 Chr.7: 81706196-81706196 on Build GRCh38
rs765831517 Chr.7: 81706299-81706299 on Build GRCh38
rs138548003 Chr.7: 81706340-81706340 on Build GRCh38
rs1016842501 Chr.7: 81706440-81706440 on Build GRCh38
rs969444645 Chr.7: 81706539-81706539 on Build GRCh38
rs5745752 Chr.7: 81706144-81706144 on Build GRCh38
rs562499092 Chr.7: 81706564-81706564 on Build GRCh38
rs915404911 Chr.7: 81706191-81706192 on Build GRCh38
rs148098479 Chr.7: 81706382-81706382 on Build GRCh38
rs1026440364 Chr.7: 81706074-81706077 on Build GRCh38
rs5745753 Chr.7: 81706122-81706122 on Build GRCh38
rs982076999 Chr.7: 81706195-81706195 on Build GRCh38
rs751689885 Chr.7: 81706331-81706331 on Build GRCh38
rs893517246 Chr.7: 81706279-81706279 on Build GRCh38
rs1025703858 Chr.7: 81706096-81706096 on Build GRCh38
rs567236033 Chr.7: 81706141-81706141 on Build GRCh38

View Full Product Details