The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_017024513.1 | XP_016880002.1 | 1 - 2 | 352 | 114 |
XM_017024516.1 | XP_016880005.1 | 1 - 2 | 140 | 114 | |
GenBank mRNA | BC037889.2 | - | 1 - 2 | 151 | 114 |
BC045824.1 | - | 1 - 2 | 151 | 114 | |
BX647411.1 | - | 1 - 2 | 157 | 114 |