The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286205.1 | NP_001273134.1 | 5 - 6 | 791 | 108 |
NM_001286209.1 | NP_001273138.1 | 5 - 6 | 683 | 108 | |
NM_001481.2 | NP_001472.1 | 5 - 6 | 668 | 108 | |
XM_005256304.4 | XP_005256361.1 | 6 - 7 | 1323 | 108 | |
XM_006721175.3 | XP_006721238.1 | 5 - 6 | 721 | 108 | |
XM_011522990.2 | XP_011521292.1 | 5 - 6 | 742 | 108 | |
XM_011522991.1 | XP_011521293.1 | 6 - 7 | 897 | 108 | |
XM_011522992.2 | XP_011521294.1 | 4 - 5 | 690 | 108 | |
XM_017023122.1 | XP_016878611.1 | 7 - 8 | 1048 | 108 | |
XM_017023123.1 | XP_016878612.1 | 5 - 6 | 747 | 108 | |
GenBank mRNA | AF050079.1 | - | 5 - 6 | 667 | 108 |
AK293958.1 | - | 5 - 6 | 698 | 108 | |
AK294050.1 | - | 4 - 5 | 517 | 108 | |
AK297857.1 | - | 5 - 6 | 651 | 108 | |
AK303505.1 | - | 5 - 6 | 724 | 108 | |
AK315261.1 | - | 5 - 6 | 616 | 108 | |
BC036781.1 | - | 6 - 7 | 108 | ||
BC104785.1 | - | 5 - 6 | 653 | 108 | |
BC112121.1 | - | 5 - 6 | 653 | 108 | |
CR749499.1 | - | 4 - 5 | 701 | 108 |