Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100408340-100408775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZCWPW1
Entrez Gene ID
55063
Gene Name
zinc finger CW-type and PWWP domain containing 1
Gene Aliases
ZCW1
UniGene
Hs.105191
Gene Chromosome Location
Chr.7: 100400826-100429239 on Build GRCh38
Transcripts
NM_001258008.1 NM_017984.4 XM_005250479.2 XM_005250480.2 XM_006716035.3 XM_006716036.2 XM_006716037.3 XM_006716038.3 XM_006716040.3 XM_017012379.1 XM_017012380.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6242506 c.884G>A p.R295H Chr.7: 100408644-100408644 on Build GRCh38
3080144 c.915G>A p.G305G Chr.7: 100408613-100408613 on Build GRCh38
3080143 c.960C>T p.I320I Chr.7: 100408568-100408568 on Build GRCh38
5405993 c.889G>A p.D297N Chr.7: 100408639-100408639 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs997113749 Chr.7: 100408713-100408713 on Build GRCh38
rs759243395 Chr.7: 100408549-100408549 on Build GRCh38
rs750747244 Chr.7: 100408740-100408740 on Build GRCh38
rs370501634 Chr.7: 100408589-100408589 on Build GRCh38
rs1028638986 Chr.7: 100408731-100408731 on Build GRCh38
rs751875514 Chr.7: 100408491-100408491 on Build GRCh38
rs774953174 Chr.7: 100408652-100408652 on Build GRCh38
rs1024780597 Chr.7: 100408447-100408448 on Build GRCh38
rs752837550 Chr.7: 100408604-100408604 on Build GRCh38
rs748172980 Chr.7: 100408509-100408509 on Build GRCh38
rs571514541 Chr.7: 100408749-100408749 on Build GRCh38
rs750233874 Chr.7: 100408595-100408595 on Build GRCh38
rs371389350 Chr.7: 100408700-100408700 on Build GRCh38
rs764112012 Chr.7: 100408562-100408562 on Build GRCh38
rs199568300 Chr.7: 100408566-100408566 on Build GRCh38
rs373658826 Chr.7: 100408644-100408644 on Build GRCh38
rs755319032 Chr.7: 100408599-100408599 on Build GRCh38
rs751281887 Chr.7: 100408512-100408512 on Build GRCh38
rs761782244 Chr.7: 100408586-100408586 on Build GRCh38
rs755232968 Chr.7: 100408499-100408499 on Build GRCh38
rs775065135 Chr.7: 100408550-100408550 on Build GRCh38
rs771324145 Chr.7: 100408539-100408539 on Build GRCh38
rs566250820 Chr.7: 100408412-100408412 on Build GRCh38
rs776187410 Chr.7: 100408671-100408671 on Build GRCh38
rs1054690203 Chr.7: 100408598-100408598 on Build GRCh38
rs538516502 Chr.7: 100408624-100408624 on Build GRCh38
rs1000728752 Chr.7: 100408395-100408395 on Build GRCh38
rs746161010 Chr.7: 100408650-100408650 on Build GRCh38
rs975787526 Chr.7: 100408704-100408709 on Build GRCh38
rs773146025 Chr.7: 100408688-100408688 on Build GRCh38
rs73401485 Chr.7: 100408673-100408673 on Build GRCh38
rs781499209 Chr.7: 100408603-100408603 on Build GRCh38
rs777877742 Chr.7: 100408622-100408622 on Build GRCh38
rs761312904 Chr.7: 100408609-100408609 on Build GRCh38
rs909410251 Chr.7: 100408384-100408384 on Build GRCh38
rs767927507 Chr.7: 100408657-100408657 on Build GRCh38
rs1019954921 Chr.7: 100408689-100408689 on Build GRCh38
rs192093712 Chr.7: 100408655-100408655 on Build GRCh38
rs548313219 Chr.7: 100408501-100408501 on Build GRCh38
rs774676030 Chr.7: 100408545-100408545 on Build GRCh38
rs750338739 Chr.7: 100408440-100408440 on Build GRCh38
rs1055004588 Chr.7: 100408402-100408402 on Build GRCh38
rs753680764 Chr.7: 100408583-100408583 on Build GRCh38
rs559084895 Chr.7: 100408715-100408715 on Build GRCh38
rs765261922 Chr.7: 100408680-100408680 on Build GRCh38
rs547921066 Chr.7: 100408382-100408382 on Build GRCh38
rs940762696 Chr.7: 100408390-100408390 on Build GRCh38
rs1045109545 Chr.7: 100408453-100408453 on Build GRCh38
rs756100160 Chr.7: 100408620-100408620 on Build GRCh38
rs779313389 Chr.7: 100408645-100408645 on Build GRCh38
rs762780467 Chr.7: 100408697-100408697 on Build GRCh38
rs1012875536 Chr.7: 100408445-100408445 on Build GRCh38
rs749693222 Chr.7: 100408538-100408538 on Build GRCh38
rs536914848 Chr.7: 100408416-100408416 on Build GRCh38

View Full Product Details