The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175921.5 | NP_787117.3 | 1 - 2 | 122 | 64 |
XM_005248289.4 | XP_005248346.1 | 1 - 2 | 153 | 64 | |
GenBank mRNA | BC131721.1 | - | 1 - 2 | 98 | 64 |
BF439305.1 | - | 1 - 2 | 86 | 64 | |
HY116483.1 | - | 1 - 2 | 123 | 64 |