Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR17
Entrez Gene ID
116966
Gene Name
WD repeat domain 17
Gene Aliases
-
Chromosome Location
Chr.4: 176065834 - 176182828 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170710.4 NP_733828.2 18 - 19 2829 97
NM_181265.3 NP_851782.3 17 - 18 2706 97
XM_005262725.3 XP_005262782.1 16 - 17 2352 97
XM_011531582.2 XP_011529884.1 17 - 18 2694 97
XM_017007686.1 XP_016863175.1 17 - 18 2682 97
XM_017007687.1 XP_016863176.1 18 - 19 2767 97
XM_017007688.1 XP_016863177.1 18 - 19 2684 97
XM_017007689.1 XP_016863178.1 17 - 18 2681 97
XM_017007690.1 XP_016863179.1 18 - 19 2767 97
XM_017007691.1 XP_016863180.1 16 - 17 2516 97
XM_017007692.1 XP_016863181.1 17 - 18 2602 97
XM_017007693.1 XP_016863182.1 16 - 17 2516 97
XM_017007694.1 XP_016863183.1 17 - 18 2500 97
XM_017007695.1 XP_016863184.1 16 - 17 2530 97
XM_017007696.1 XP_016863185.1 16 - 17 2530 97
XM_017007697.1 XP_016863186.1 17 - 18 2683 97
GenBank mRNA AF492460.1 - 18 - 19 2682 97
AK130128.1 - 3 - 4 336 97
BX648508.1 - 2 - 3 253 97

View Full Product Details