The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006855.3 | NP_006846.1 | 3 - 4 | 526 | 91 |
NM_016657.2 | NP_057839.1 | 3 - 4 | 526 | 91 | |
GenBank mRNA | AK129888.1 | - | 3 - 4 | 492 | 91 |
AK222930.1 | - | 3 - 4 | 506 | 91 | |
AK223238.1 | - | 3 - 4 | 483 | 91 | |
AK292102.1 | - | 3 - 4 | 518 | 91 | |
AL035081.1 | - | 3 - 4 | 479 | 91 | |
AL035082.1 | - | 3 - 4 | 510 | 91 | |
BC001277.1 | - | 3 - 4 | 503 | 91 | |
BC096722.1 | - | 3 - 4 | 460 | 91 | |
BC098131.1 | - | 3 - 4 | 460 | 91 | |
BC098156.1 | - | 3 - 4 | 460 | 91 | |
BC098336.1 | - | 3 - 4 | 460 | 91 | |
BU621350.1 | - | 1 - 2 | 91 | ||
CR456509.1 | - | 3 - 4 | 401 | 91 |