Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1361690-1362287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNPTG
Entrez Gene ID
84572
Gene Name
N-acetylglucosamine-1-phosphate transferase gamma subunit
Gene Aliases
C16orf27, GNPTAG, LP2537, RJD9
UniGene
Hs.241575
Gene Chromosome Location
Chr.16: 1351899-1363351 on Build GRCh38
Transcripts
NM_032520.4 XM_017023782.1 XM_017023783.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6672465 c.485C>T p.T162M Chr.16: 1362279-1362279 on Build GRCh38
4255909 c.211T>C p.C71R Chr.16: 1361775-1361775 on Build GRCh38
1253377 c.425G>A p.C142Y Chr.16: 1362219-1362219 on Build GRCh38
6672466 c.183C>T p.P61P Chr.16: 1361747-1361747 on Build GRCh38
1208435 c.340G>A p.A114T Chr.16: 1362060-1362060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778178434 Chr.16: 1361889-1361889 on Build GRCh38
rs201531462 Chr.16: 1361915-1361915 on Build GRCh38
rs1032142138 Chr.16: 1362010-1362010 on Build GRCh38
rs764395288 Chr.16: 1361903-1361903 on Build GRCh38
rs201992413 Chr.16: 1361914-1361914 on Build GRCh38
rs562132501 Chr.16: 1362242-1362242 on Build GRCh38
rs149197775 Chr.16: 1361907-1361907 on Build GRCh38
rs901500768 Chr.16: 1361996-1361996 on Build GRCh38
rs564859061 Chr.16: 1362265-1362265 on Build GRCh38
rs765847099 Chr.16: 1361923-1361923 on Build GRCh38
rs772637099 Chr.16: 1361795-1361795 on Build GRCh38
rs772275436 Chr.16: 1361840-1361840 on Build GRCh38
rs756379978 Chr.16: 1362216-1362216 on Build GRCh38
rs374890181 Chr.16: 1362234-1362234 on Build GRCh38
rs148378749 Chr.16: 1362020-1362020 on Build GRCh38
rs753100529 Chr.16: 1362159-1362160 on Build GRCh38
rs112850896 Chr.16: 1361871-1361871 on Build GRCh38
rs754010312 Chr.16: 1361755-1361755 on Build GRCh38
rs769722097 Chr.16: 1362045-1362045 on Build GRCh38
rs757509701 Chr.16: 1362144-1362144 on Build GRCh38
rs758065510 Chr.16: 1361821-1361821 on Build GRCh38
rs201978812 Chr.16: 1362005-1362005 on Build GRCh38
rs745552342 Chr.16: 1361775-1361775 on Build GRCh38
rs750858322 Chr.16: 1362172-1362172 on Build GRCh38
rs759479821 Chr.16: 1361855-1361855 on Build GRCh38
rs765865746 Chr.16: 1362118-1362118 on Build GRCh38
rs779191841 Chr.16: 1362025-1362025 on Build GRCh38
rs779778381 Chr.16: 1361880-1361880 on Build GRCh38
rs753679575 Chr.16: 1362254-1362254 on Build GRCh38
rs768086598 Chr.16: 1362133-1362134 on Build GRCh38
rs199782632 Chr.16: 1361808-1361808 on Build GRCh38
rs752300611 Chr.16: 1361969-1361969 on Build GRCh38
rs371836175 Chr.16: 1361842-1361842 on Build GRCh38
rs781036850 Chr.16: 1362092-1362092 on Build GRCh38
rs776359831 Chr.16: 1361721-1361721 on Build GRCh38
rs138018487 Chr.16: 1362266-1362266 on Build GRCh38
rs367661016 Chr.16: 1362060-1362060 on Build GRCh38
rs940572438 Chr.16: 1362221-1362221 on Build GRCh38
rs368718738 Chr.16: 1362193-1362193 on Build GRCh38
rs774546219 Chr.16: 1361731-1361731 on Build GRCh38
rs757226714 Chr.16: 1361826-1361826 on Build GRCh38
rs376636252 Chr.16: 1361833-1361833 on Build GRCh38
rs758027840 Chr.16: 1361970-1361970 on Build GRCh38
rs780912876 Chr.16: 1361830-1361830 on Build GRCh38
rs375386449 Chr.16: 1362146-1362146 on Build GRCh38
rs781337088 Chr.16: 1362145-1362145 on Build GRCh38
rs780619430 Chr.16: 1362256-1362256 on Build GRCh38
rs745994739 Chr.16: 1361897-1361897 on Build GRCh38
rs761584357 Chr.16: 1362108-1362108 on Build GRCh38
rs777411480 Chr.16: 1361714-1361714 on Build GRCh38
rs375764145 Chr.16: 1362026-1362026 on Build GRCh38
rs184790515 Chr.16: 1361992-1361992 on Build GRCh38
rs73487805 Chr.16: 1362189-1362189 on Build GRCh38
rs767895135 Chr.16: 1361744-1361744 on Build GRCh38
rs763669202 Chr.16: 1362176-1362183 on Build GRCh38
rs143316080 Chr.16: 1362019-1362019 on Build GRCh38
rs760008789 Chr.16: 1362171-1362171 on Build GRCh38
rs774017237 Chr.16: 1362051-1362051 on Build GRCh38
rs9926432 Chr.16: 1361804-1361804 on Build GRCh38
rs747362604 Chr.16: 1361892-1361892 on Build GRCh38
rs900341724 Chr.16: 1361997-1361997 on Build GRCh38
rs1000273462 Chr.16: 1362262-1362262 on Build GRCh38
rs113388753 Chr.16: 1362258-1362258 on Build GRCh38
rs754679713 Chr.16: 1362255-1362255 on Build GRCh38
rs371376391 Chr.16: 1362007-1362007 on Build GRCh38
rs559549488 Chr.16: 1361968-1361968 on Build GRCh38
rs767760180 Chr.16: 1362183-1362183 on Build GRCh38
rs200975355 Chr.16: 1362067-1362067 on Build GRCh38
rs767633692 Chr.16: 1362247-1362247 on Build GRCh38
rs774835763 Chr.16: 1361783-1361783 on Build GRCh38
rs281864956 Chr.16: 1362239-1362239 on Build GRCh38
rs371413637 Chr.16: 1361826-1361827 on Build GRCh38
rs760265920 Chr.16: 1362172-1362173 on Build GRCh38
rs773268405 Chr.16: 1362249-1362249 on Build GRCh38
rs757318601 Chr.16: 1361869-1361869 on Build GRCh38
rs763430254 Chr.16: 1361725-1361725 on Build GRCh38
rs369333639 Chr.16: 1361864-1361864 on Build GRCh38
rs200741370 Chr.16: 1362114-1362114 on Build GRCh38
rs770716391 Chr.16: 1362038-1362038 on Build GRCh38
rs200116947 Chr.16: 1361991-1361991 on Build GRCh38
rs973297006 Chr.16: 1361724-1361724 on Build GRCh38
rs762024309 Chr.16: 1361787-1361787 on Build GRCh38
rs749320662 Chr.16: 1361942-1361942 on Build GRCh38
rs745850911 Chr.16: 1362043-1362043 on Build GRCh38
rs201045559 Chr.16: 1361935-1361935 on Build GRCh38
rs760954235 Chr.16: 1362250-1362250 on Build GRCh38
rs201381376 Chr.16: 1361841-1361841 on Build GRCh38
rs768465491 Chr.16: 1362153-1362153 on Build GRCh38
rs771330629 Chr.16: 1361988-1361992 on Build GRCh38
rs775359476 Chr.16: 1362096-1362096 on Build GRCh38
rs193302849 Chr.16: 1362067-1362069 on Build GRCh38
rs555838948 Chr.16: 1361818-1361818 on Build GRCh38
rs371195304 Chr.16: 1361958-1361958 on Build GRCh38
rs139385234 Chr.16: 1361940-1361940 on Build GRCh38
rs756455159 Chr.16: 1362016-1362016 on Build GRCh38
rs777887031 Chr.16: 1362081-1362081 on Build GRCh38
rs775985395 Chr.16: 1362233-1362233 on Build GRCh38
rs76594024 Chr.16: 1361893-1361893 on Build GRCh38
rs552402857 Chr.16: 1361767-1361767 on Build GRCh38
rs543804571 Chr.16: 1361908-1361908 on Build GRCh38
rs201566045 Chr.16: 1361800-1361800 on Build GRCh38
rs373810840 Chr.16: 1361816-1361816 on Build GRCh38
rs763127645 Chr.16: 1361926-1361926 on Build GRCh38
rs906630870 Chr.16: 1362160-1362160 on Build GRCh38
rs2072987 Chr.16: 1362004-1362004 on Build GRCh38
rs774898087 Chr.16: 1362167-1362167 on Build GRCh38
rs778810991 Chr.16: 1361761-1361761 on Build GRCh38
rs767502955 Chr.16: 1361873-1361873 on Build GRCh38
rs777517766 Chr.16: 1361768-1361768 on Build GRCh38
rs140264422 Chr.16: 1361747-1361747 on Build GRCh38
rs143383133 Chr.16: 1361933-1361933 on Build GRCh38
rs367754651 Chr.16: 1361983-1361983 on Build GRCh38
rs952698680 Chr.16: 1361867-1361874 on Build GRCh38
rs748576549 Chr.16: 1361838-1361838 on Build GRCh38
rs779795759 Chr.16: 1362149-1362149 on Build GRCh38
rs766619396 Chr.16: 1361750-1361750 on Build GRCh38
rs775693232 Chr.16: 1362175-1362175 on Build GRCh38
rs147048228 Chr.16: 1362059-1362059 on Build GRCh38
rs762446439 Chr.16: 1361732-1361732 on Build GRCh38
rs747291812 Chr.16: 1362082-1362082 on Build GRCh38
rs193302848 Chr.16: 1361760-1361760 on Build GRCh38
rs763065729 Chr.16: 1362251-1362251 on Build GRCh38
rs773452586 Chr.16: 1361876-1361881 on Build GRCh38
rs200915537 Chr.16: 1361852-1361852 on Build GRCh38
rs200362559 Chr.16: 1361805-1361805 on Build GRCh38
rs376327160 Chr.16: 1361867-1361867 on Build GRCh38
rs745356243 Chr.16: 1361831-1361831 on Build GRCh38
rs746507546 Chr.16: 1361715-1361715 on Build GRCh38
rs372039531 Chr.16: 1361774-1361774 on Build GRCh38
rs769001628 Chr.16: 1361950-1361950 on Build GRCh38
rs753205380 Chr.16: 1362120-1362120 on Build GRCh38
rs775604993 Chr.16: 1361901-1361901 on Build GRCh38
rs758452962 Chr.16: 1362077-1362077 on Build GRCh38
rs778481117 Chr.16: 1361928-1361928 on Build GRCh38
rs778657195 Chr.16: 1362198-1362198 on Build GRCh38
rs751676968 Chr.16: 1362178-1362178 on Build GRCh38
rs751592131 Chr.16: 1361822-1361822 on Build GRCh38
rs200508759 Chr.16: 1362176-1362176 on Build GRCh38
rs748272191 Chr.16: 1361814-1361815 on Build GRCh38
rs768511134 Chr.16: 1361926-1361926 on Build GRCh38
rs761835152 Chr.16: 1362179-1362179 on Build GRCh38
rs371160615 Chr.16: 1362015-1362015 on Build GRCh38
rs374558065 Chr.16: 1361967-1361967 on Build GRCh38
rs141870165 Chr.16: 1362095-1362095 on Build GRCh38
rs770325965 Chr.16: 1361936-1361936 on Build GRCh38
rs753385216 Chr.16: 1361927-1361927 on Build GRCh38
rs1033139347 Chr.16: 1362124-1362124 on Build GRCh38
rs747713846 Chr.16: 1362200-1362200 on Build GRCh38
rs137852884 Chr.16: 1362053-1362053 on Build GRCh38
rs752553225 Chr.16: 1361817-1361817 on Build GRCh38
rs751393135 Chr.16: 1361868-1361868 on Build GRCh38
rs368983807 Chr.16: 1361876-1361876 on Build GRCh38
rs747545433 Chr.16: 1362161-1362161 on Build GRCh38
rs749298719 Chr.16: 1362151-1362151 on Build GRCh38
rs756056220 Chr.16: 1361922-1361922 on Build GRCh38
rs750277954 Chr.16: 1361917-1361917 on Build GRCh38
rs776376435 Chr.16: 1362041-1362041 on Build GRCh38
rs112358354 Chr.16: 1361976-1361976 on Build GRCh38
rs144929410 Chr.16: 1361832-1361832 on Build GRCh38
rs200507633 Chr.16: 1361797-1361797 on Build GRCh38
rs769323972 Chr.16: 1361782-1361782 on Build GRCh38
rs763335225 Chr.16: 1362125-1362125 on Build GRCh38
rs764763952 Chr.16: 1362008-1362008 on Build GRCh38
rs193302847 Chr.16: 1362037-1362037 on Build GRCh38
rs748782055 Chr.16: 1362101-1362101 on Build GRCh38
rs372004052 Chr.16: 1361814-1361814 on Build GRCh38
rs771863638 Chr.16: 1362163-1362163 on Build GRCh38
rs554418793 Chr.16: 1362109-1362109 on Build GRCh38
rs770725085 Chr.16: 1361716-1361716 on Build GRCh38
rs763678034 Chr.16: 1362048-1362048 on Build GRCh38
rs776752687 Chr.16: 1361853-1361853 on Build GRCh38
rs200003910 Chr.16: 1361866-1361866 on Build GRCh38
rs777991598 Chr.16: 1362028-1362028 on Build GRCh38
rs764279342 Chr.16: 1362128-1362128 on Build GRCh38
rs781472406 Chr.16: 1361971-1361971 on Build GRCh38
rs752836392 Chr.16: 1362070-1362070 on Build GRCh38
rs528538033 Chr.16: 1362192-1362192 on Build GRCh38
rs376026722 Chr.16: 1362009-1362009 on Build GRCh38
rs768817513 Chr.16: 1361988-1361988 on Build GRCh38
rs774673554 Chr.16: 1361953-1361953 on Build GRCh38
rs748941730 Chr.16: 1361881-1361881 on Build GRCh38
rs772949824 Chr.16: 1362170-1362170 on Build GRCh38
rs1038742616 Chr.16: 1362033-1362033 on Build GRCh38
rs769814085 Chr.16: 1361844-1361845 on Build GRCh38
rs200574942 Chr.16: 1361748-1361748 on Build GRCh38
rs769110491 Chr.16: 1361726-1361726 on Build GRCh38
rs376679416 Chr.16: 1361798-1361798 on Build GRCh38
rs754595108 Chr.16: 1361884-1361884 on Build GRCh38
rs145313679 Chr.16: 1361902-1361902 on Build GRCh38
rs769717288 Chr.16: 1361898-1361898 on Build GRCh38
rs988175540 Chr.16: 1361742-1361742 on Build GRCh38
rs372319167 Chr.16: 1362102-1362102 on Build GRCh38
rs530486639 Chr.16: 1362217-1362217 on Build GRCh38
rs370815540 Chr.16: 1361959-1361959 on Build GRCh38
rs926216364 Chr.16: 1361962-1361962 on Build GRCh38
rs749952316 Chr.16: 1361875-1361875 on Build GRCh38
rs775539912 Chr.16: 1361863-1361863 on Build GRCh38
rs751575030 Chr.16: 1362135-1362135 on Build GRCh38
rs137852885 Chr.16: 1361954-1361954 on Build GRCh38
rs757581909 Chr.16: 1361932-1361932 on Build GRCh38
rs1005797412 Chr.16: 1362185-1362185 on Build GRCh38
rs756586617 Chr.16: 1361979-1361979 on Build GRCh38
rs771424681 Chr.16: 1361846-1361846 on Build GRCh38
rs147109185 Chr.16: 1362244-1362244 on Build GRCh38
rs960073699 Chr.16: 1362013-1362013 on Build GRCh38
rs772946278 Chr.16: 1362001-1362001 on Build GRCh38
rs141533972 Chr.16: 1362047-1362047 on Build GRCh38
rs762664496 Chr.16: 1362011-1362011 on Build GRCh38
rs576804198 Chr.16: 1362154-1362154 on Build GRCh38
rs11554983 Chr.16: 1362115-1362115 on Build GRCh38
rs113058618 Chr.16: 1362014-1362014 on Build GRCh38
rs193302850 Chr.16: 1362099-1362111 on Build GRCh38
rs772324860 Chr.16: 1361995-1361995 on Build GRCh38
rs138297609 Chr.16: 1362064-1362064 on Build GRCh38
rs756307321 Chr.16: 1362190-1362190 on Build GRCh38
rs757394606 Chr.16: 1362086-1362086 on Build GRCh38
rs769233183 Chr.16: 1362235-1362235 on Build GRCh38
rs767072821 Chr.16: 1362006-1362006 on Build GRCh38
rs375115440 Chr.16: 1361856-1361856 on Build GRCh38

View Full Product Details