Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACPT
Entrez Gene ID
93650
Gene Name
acid phosphatase, testicular
Gene Aliases
-
Chromosome Location
Chr.19: 50790415 - 50795224 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033068.2 NP_149059.1 8 - 9 863 108

View Full Product Details