The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000715.3 | NP_000706.1 | 2 - 3 | 339 | 101 |
XM_005273251.1 | XP_005273308.1 | 2 - 3 | 295 | 101 | |
XM_005273252.4 | XP_005273309.1 | 2 - 3 | 471 | 101 | |
GenBank mRNA | AK313164.1 | - | 2 - 3 | 285 | 101 |
BC022312.1 | - | 2 - 3 | 339 | 101 | |
M31452.1 | - | 2 - 3 | 283 | 101 | |
X07853.1 | - | 1 - 2 | 153 | 101 |