The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000715.3 | NP_000706.1 | 7 - 8 | 1083 | 105 |
XM_005273251.1 | XP_005273308.1 | 7 - 8 | 1039 | 105 | |
XM_005273252.4 | XP_005273309.1 | 7 - 8 | 1215 | 105 | |
GenBank mRNA | AK303793.1 | - | 8 - 9 | 105 | |
AK303798.1 | - | 7 - 8 | 1033 | 105 | |
AK313164.1 | - | 7 - 8 | 1029 | 105 | |
BC022312.1 | - | 7 - 8 | 1083 | 105 | |
M31452.1 | - | 7 - 8 | 1027 | 105 | |
X02865.1 | - | 5 - 6 | 652 | 105 |