The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001043.3 | NP_001034.1 | 3 - 4 | 899 | 86 |
NM_001172501.1 | NP_001165972.1 | 4 - 5 | 1235 | 86 | |
NM_001172502.1 | NP_001165973.1 | 1 - 2 | 505 | 86 | |
NM_001172504.1 | NP_001165975.1 | 3 - 4 | 695 | 86 | |
XM_006721263.2 | XP_006721326.1 | 4 - 5 | 794 | 86 | |
XM_011523295.1 | XP_011521597.1 | 4 - 5 | 1261 | 86 | |
XM_011523296.1 | XP_011521598.1 | 4 - 5 | 1261 | 86 | |
XM_011523297.2 | XP_011521599.1 | 4 - 5 | 1261 | 86 | |
XM_011523298.1 | XP_011521600.1 | 4 - 5 | 1261 | 86 | |
XM_011523299.2 | XP_011521601.1 | 2 - 3 | 382 | 86 | |
XM_011523300.2 | XP_011521602.1 | 2 - 3 | 1465 | 86 | |
GenBank mRNA | AK301811.1 | - | 1 - 2 | 465 | 86 |
AK312793.1 | - | 4 - 5 | 729 | 86 | |
BC060831.1 | - | 3 - 4 | 818 | 86 | |
M65105.1 | - | 3 - 4 | 704 | 86 |